This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02452285_g1
See other UNC-49 GE Assays ›
Gene Symbol:
unc-49
Gene Name:
protein-coding
Gene Aliases:
CELE_T21C12.1
Chromosome Location:
Chr.III: 10520762 - 10532706 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02452285_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: unc-49
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001027611.3 NP_001022782.1 21 - 22 2739 58
NM_001027612.4 NP_001022783.1 14 - 15 1922 58
NM_001027613.3 NP_001022784.1 13 - 14 1886 58
NM_001027614.4 NP_001022785.1 15 - 16 1898 58
NM_001027615.4 NP_001022786.1 7 - 8 1021 58
NM_001306734.3 NP_001293663.1 3 - 4 311 58
NM_001379901.2 NP_001368252.1 13 - 14 2469 58
NM_001379904.1 NP_001367142.1 13 - 14 2550 58
GenBank mRNA AF151641.1 - 14 - 15 1848 58
AF151642.1 - 13 - 14 1812 58
AF151643.1 - 15 - 16 1905 58
AF151644.1 - 7 - 8 928 58
AF151645.1 - 2 - 3 427 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ion transport
signal transduction
gamma-aminobutyric acid signaling pathway
chemical synaptic transmission
locomotory behavior
ion transmembrane transport
regulation of membrane potential
regulation of backward locomotion
neurological system process
chloride transmembrane transport
transmembrane signaling receptor activity
GABA-A receptor activity
ion channel activity
extracellular ligand-gated ion channel activity
chloride channel activity
protein binding
neurotransmitter receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0