This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02449113_m1
See other FBN-1 GE Assays ›
Gene Symbol:
fbn-1
Gene Name:
protein-coding
Gene Aliases:
CELE_ZK783.1
Chromosome Location:
Chr.III: 7625376 - 7641078 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02449113_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: fbn-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001276783.3 NP_001263712.1 14 - 15 5845 111
NM_001276785.3 NP_001263714.1 14 - 15 5845 111
NM_066269.6 NP_498670.2 14 - 15 5845 111

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Process(es)

calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0