This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02444183_m1
See other LAF-1 GE Assays ›
Gene Symbol:
laf-1
Gene Name:
ATP-dependent RNA helicase laf-1
Gene Aliases:
CELE_Y71H2AM.19
Chromosome Location:
Chr.III: 2768865 - 2779053 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02444183_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: laf-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001267930.3 NP_001254859.1 4 - 5 1651 143
GenBank mRNA FJ348231.1 - 4 - 5 1673 143

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of translation
gamete generation
negative regulation of gene expression
cell differentiation
positive regulation of embryonic development
masculinization of hermaphroditic germ-line
positive regulation of fertilization
nucleotide binding
nucleic acid binding
RNA binding
RNA helicase activity
helicase activity
ATP binding
RNA-dependent ATPase activity
hydrolase activity
RNA strand annealing activity
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0