This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02435650_g1
See other IRE-1 GE Assays ›
Gene Symbol:
ire-1
Gene Name:
protein-coding
Gene Aliases:
CELE_C41C4.4
Chromosome Location:
Chr.II: 8112243 - 8126876 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02435650_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ire-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001267206.3 NP_001254135.1 8 - 9 2389 122
NM_001267207.3 NP_001254136.1 2 - 3 1214 122
GenBank mRNA AF435952.1 - 7 - 8 2252 122

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
nematode larval development
mRNA processing
protein phosphorylation
response to unfolded protein
positive regulation of transcription from RNA polymerase II promoter involved in unfolded protein response
age-dependent general metabolic decline
metabolic process
phosphorylation
endoplasmic reticulum unfolded protein response
response to topologically incorrect protein
IRE1-mediated unfolded protein response
negative regulation of developmental process
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
nucleic acid phosphodiester bond hydrolysis
RNA phosphodiester bond hydrolysis
RNA phosphodiester bond hydrolysis, endonucleolytic
positive regulation of intrinsic apoptotic signaling pathway by p53 class mediator
response to tunicamycin
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
nucleotide binding
catalytic activity
endoribonuclease activity
ribonuclease activity
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
hydrolase activity
unfolded protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0