This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02433464_g1
See other PHB-2 GE Assays ›
Gene Symbol:
phb-2
Gene Name:
Mitochondrial prohibitin complex protein 2
Gene Aliases:
CELE_T24H7.1
Chromosome Location:
Chr.II: 6251411 - 6252780 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02433464_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: phb-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_062849.5 NP_495250.2 3 - 4 474 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s)

regulation of oxidative phosphorylation
response to oxidative stress
spermatogenesis
gonad development
embryo development ending in birth or egg hatching
defecation
positive regulation of multicellular organism growth
regulation of pharyngeal pumping
oogenesis
mitochondrion morphogenesis

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0