This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02433337_m1
See other RGS-3 GE Assays ›
Gene Symbol:
rgs-3
Gene Name:
protein-coding
Gene Aliases:
CELE_C29H12.3
Chromosome Location:
Chr.II: 6099800 - 6110126 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02433337_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rgs-3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_062822.4 NP_495223.1 3 - 4 450 73
NM_182096.6 NP_871896.1 2 - 3 669 73

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_182096.6 transcripts.

Set Membership:

Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s)

chemosensory behavior
negative regulation of signal transduction
sensory perception of bitter taste

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header