This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02430228_m1
See other SMU-2 GE Assays ›
Gene Symbol:
smu-2
Gene Name:
Smu-2 suppressor of mec-8 and unc-52 protein
Gene Aliases:
CELE_Y49F6B.4
Chromosome Location:
Chr.II: 3482408 - 3490151 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02430228_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: smu-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_062158.5 NP_494559.1 6 - 7 1546 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
nematode larval development
mRNA processing
mechanosensory behavior
RNA splicing
embryo development ending in birth or egg hatching
locomotion
muscle organ morphogenesis
neuron development
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0