This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02426924_m1
See other NFYB-1 GE Assays ›
Gene Symbol:
nfyb-1
Gene Name:
protein-coding
Gene Aliases:
CELE_W10D9.4
Chromosome Location:
Chr.II: 461664 - 467365 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02426924_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: nfyb-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001316872.3 NP_001303801.1 2 - 3 342 118
NM_061339.6 NP_493740.1 3 - 4 593 118

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0