This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02426642_m1
See other ETR-1 GE Assays ›
Gene Symbol:
etr-1
Gene Name:
protein-coding
Gene Aliases:
CELE_T01D1.2
Chromosome Location:
Chr.II: 154166 - 170630 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02426642_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: etr-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001381290.2 NP_001367867.1 1 - 2 243 76
NM_001381291.1 NP_001368521.1 1 - 2 222 76
NM_001381292.2 NP_001367871.1 2 - 3 223 76
NM_001381293.1 NP_001367870.1 2 - 3 334 76
NM_001381294.1 NP_001367872.1 2 - 3 431 76
NM_001383771.1 1 - 2 181 76
NR_132573.1 1 - 2 181 76
GenBank mRNA EF523766.1 - 1 - 2 181 76
EF523770.1 - 1 - 2 181 76
U53931.1 - 1 - 2 244 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
mRNA splice site selection
determination of adult lifespan
nucleic acid binding
RNA binding
mRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0