This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02423363_m1
See other DYS-1 GE Assays ›
Gene Symbol:
dys-1
Gene Name:
protein-coding
Gene Aliases:
CELE_F15D3.1
Chromosome Location:
Chr.I: 11526918 - 11557854 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02423363_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: dys-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001306310.3 NP_001293239.1 25 - 26 6402 61
NM_001306311.3 NP_001293240.1 14 - 15 3603 61
NM_001306312.3 NP_001293241.1 2 - 3 789 61
NM_001306313.4 NP_001293242.1 28 - 29 7047 61
NM_001306314.3 NP_001293243.1 24 - 25 6369 61
NM_001383731.1 24 - 25 6420 61
NM_060545.6 NP_492946.1 29 - 30 7142 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

synaptic transmission, cholinergic
locomotory behavior
acetylcholine transport
positive regulation of synaptic transmission, cholinergic
positive regulation of locomotion
forward locomotion
sarcomere organization
muscle cell cellular homeostasis
synaptic signaling
actin binding
protein binding
zinc ion binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0