This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02422414_m1
See other UNC-82 GE Assays ›
Gene Symbol:
unc-82
Gene Name:
protein-coding
Gene Aliases:
CELE_B0496.3
Chromosome Location:
Chr.IV: 7424498 - 7444664 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02422414_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: unc-82
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001268365.3 NP_001255294.1 25 - 26 4233 91
NM_001268366.3 NP_001255295.1 20 - 21 3439 91
NM_001268369.2 NP_001255298.1 21 - 22 3427 91
NM_001307278.3 NP_001294207.1 21 - 22 3637 91
NM_001372869.1 NP_001359805.1 21 - 22 3427 91
NM_001380324.1 NP_001367366.1 22 - 23 3619 91
NM_001380325.1 NP_001367367.1 24 - 25 3860 91
NM_001380326.1 NP_001367365.1 2 - 3 221 91
NM_068785.7 NP_501186.5 21 - 22 3586 91

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001380326.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
phosphorylation
intracellular signal transduction
cellular response to glucose starvation
striated muscle paramyosin thick filament assembly
striated muscle myosin thick filament assembly
protein kinase activity
protein serine/threonine kinase activity
ATP binding
structural constituent of muscle
kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0