This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02421675_m1
See other DIN-1 GE Assays ›
Gene Symbol:
din-1
Gene Name:
protein-coding
Gene Aliases:
CELE_F07A11.6
Chromosome Location:
Chr.II: 11614624 - 11633611 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02421675_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: din-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001267377.3 NP_001254306.1 11 - 12 5293 70
NM_001381614.1 NP_001367110.1 17 - 18 6449 70
NM_064083.6 NP_496484.3 15 - 16 5907 70
NM_064084.8 NP_496485.2 16 - 17 6497 70
NM_182101.5 NP_871901.1 17 - 18 6427 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
regulation of transcription, DNA-templated
transcription regulatory region sequence-specific DNA binding
nucleic acid binding
RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0