This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02420610_g1
See other SMA-9 GE Assays ›
Gene Symbol:
sma-9
Gene Name:
protein-coding
Gene Aliases:
CELE_T05A10.1
Chromosome Location:
Chr.X: 10769282 - 10782973 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02420610_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: sma-9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001375119.3 NP_001361905.1 6 - 7 1886 93
NM_001375120.2 NP_001361887.1 7 - 8 2333 93
NM_001375121.3 NP_001361888.1 7 - 8 2333 93
NM_001375122.2 NP_001361889.1 7 - 8 2333 93
NM_001375123.1 NP_001361886.1 7 - 8 2333 93
NM_001375124.1 NP_001361885.1 8 - 9 2414 93
NM_001375125.2 NP_001361884.1 6 - 7 1880 93
NM_001377620.1 NP_001364832.1 7 - 8 2009 93
NM_001383639.1 7 - 8 2009 93
NM_171771.6 NP_741896.2 5 - 6 1805 93

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001375125.2 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transforming growth factor beta receptor signaling pathway
positive regulation of multicellular organism growth
regulation of mesodermal cell fate specification
nematode male tail tip morphogenesis
positive regulation of mesodermal cell fate specification
negative regulation of nucleic acid-templated transcription
positive regulation of nucleic acid-templated transcription
nucleic acid binding
transcription coactivator activity
transcription corepressor activity
zinc ion binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0