This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02419739_m1
See other UNC-2 GE Assays ›
Gene Symbol:
unc-2
Gene Name:
protein-coding
Gene Aliases:
CELE_T02C5.5
Chromosome Location:
Chr.X: 2710920 - 2763526 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02419739_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: unc-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001348588.4 NP_001335561.1 17 - 18 3578 81
NM_001348589.4 NP_001335563.1 17 - 18 3578 81
NM_001348590.4 NP_001335560.1 17 - 18 3578 81
NM_001348591.4 NP_001335562.1 17 - 18 3578 81
NM_001348592.5 NP_001335571.1 17 - 18 3773 81
NM_001348593.4 NP_001335573.1 17 - 18 3773 81
NM_001348594.5 NP_001335570.1 17 - 18 3773 81
NM_001348595.4 NP_001335572.1 17 - 18 3773 81
NM_001348596.5 NP_001335516.1 15 - 16 3305 81
NM_001348597.2 NP_001335515.1 15 - 16 3305 81
NM_001368494.4 NP_001355464.1 23 - 24 4139 81
NM_001368495.4 NP_001355463.1 21 - 22 3983 81
NM_001368496.4 NP_001355462.1 21 - 22 4007 81
NM_001368497.4 NP_001355494.1 17 - 18 3906 81
NM_171638.3 NP_741732.2 17 - 18 3763 81
GenBank mRNA AY264781.1 - 17 - 18 3763 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of neurotransmitter levels
ion transport
calcium ion transport
chemical synaptic transmission
locomotory behavior
response to heat
negative regulation of transforming growth factor beta receptor signaling pathway
regulation of ion transmembrane transport
determination of left/right asymmetry in nervous system
response to starvation
transmembrane transport
calcium ion import
calcium ion transmembrane transport
ion channel activity
voltage-gated ion channel activity
voltage-gated calcium channel activity
calcium channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0