This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02419623_m1
See other NCAM-1 GE Assays ›
Gene Symbol:
ncam-1
Gene Name:
protein-coding
Gene Aliases:
CELE_F02G3.1
Chromosome Location:
Chr.X: 691913 - 703040 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02419623_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ncam-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_171615.7 NP_741707.3 1 - 2 1167 99
NM_171616.5 NP_741708.1 1 - 2 51 99
GenBank mRNA AF456360.2 - 1 - 2 75 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
axon guidance
dendrite self-avoidance
protein binding involved in cell-cell adhesion

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0