This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02417353_g1
See other MTM-1 GE Assays ›
Gene Symbol:
mtm-1
Gene Name:
Myotubularin-related protein 1
Gene Aliases:
CELE_Y110A7A.5
Chromosome Location:
Chr.I: 5134709 - 5137971 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02417353_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mtm-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_059130.5 NP_491531.2 1 - 2 100 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
phagocytosis
peptidyl-tyrosine dephosphorylation
negative regulation of endocytosis
phospholipid dephosphorylation
phosphatidylinositol dephosphorylation
negative regulation of engulfment of apoptotic cell
phosphatidylinositol-3-phosphatase activity
protein binding
phosphatidylinositol-4,5-bisphosphate binding
hydrolase activity
phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0