This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02416857_m1
See other CHD-7 GE Assays ›
Gene Symbol:
chd-7
Gene Name:
Chromodomain and Helicase Domain protein
Gene Aliases:
CELE_T04D1.4
Chromosome Location:
Chr.I: 4685863 - 4698575 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02416857_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: chd-7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_059025.7 NP_491426.2 5 - 6 3786 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Process(es)

helicase activity
ATP binding
hydrolase activity, acting on acid anhydrides

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0