This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02415115_g1
See other COPA-1 GE Assays ›
Gene Symbol:
copa-1
Gene Name:
Coatomer subunit alpha
Gene Aliases:
CELE_Y71F9AL.17
Chromosome Location:
Chr.I: 2895329 - 2900860 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02415115_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: copa-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_058668.6 NP_491069.1 1 - 2 99 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
ER to Golgi vesicle-mediated transport
retrograde vesicle-mediated transport, Golgi to ER
intra-Golgi vesicle-mediated transport
protein transport
vesicle-mediated transport
structural molecule activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0