This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02414654_m1
See other ABT-2 GE Assays ›
Gene Symbol:
abt-2
Gene Name:
protein-coding
Gene Aliases:
CELE_F12B6.1
Chromosome Location:
Chr.I: 2255921 - 2272866 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02414654_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: abt-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001381367.1 NP_001368466.1 32 - 33 6145 60
NM_058548.7 NP_490949.3 32 - 33 6256 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

lipid transport
transmembrane transport
nucleotide binding
lipid transporter activity
ATP binding
ATPase activity
ATPase activity, coupled to transmembrane movement of substances

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0