This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02410960_g1
See other GON-2 GE Assays ›
Gene Symbol:
gon-2
Gene Name:
protein-coding
Gene Aliases:
CELE_T01H8.5
Chromosome Location:
Chr.I: 8549510 - 8570998 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02410960_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gon-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001129034.3 NP_001122506.1 11 - 12 2183 129
NM_001136316.4 NP_001129788.1 5 - 6 1148 129
NM_001373178.2 NP_001359707.1 13 - 14 2334 129
NM_059914.7 NP_492315.4 14 - 15 2407 129

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mitotic cell cycle
ion transport
cation transport
gonad development
defecation
ion transmembrane transport
transmembrane transport
divalent metal ion transport
cation transmembrane transport
ion channel activity
cation channel activity
cation transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0