This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02406334_m1
See other PAT-12 GE Assays ›
Gene Symbol:
pat-12
Gene Name:
protein-coding
Gene Aliases:
CELE_T17H7.4
Chromosome Location:
Chr.III: 727207 - 752432 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02406334_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pat-12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001373834.2 NP_001359886.1 2 - 3 191 94
NM_001381698.1 NP_001367550.1 1 - 2 88 94
NM_001381699.1 NP_001367549.1 1 - 2 92 94
NM_001381700.1 NP_001367551.1 1 - 2 88 94
NM_001381704.2 NP_001367553.1 3 - 4 279 94
NM_171067.5 NP_741076.1 3 - 4 273 94

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001373834.2, NM_001381704.2 transcripts.

Set Membership:

Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

hemidesmosome assembly
embryonic morphogenesis
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0