This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02406203_m1
See other PTP-3 GE Assays ›
Gene Symbol:
ptp-3
Gene Name:
protein-coding
Gene Aliases:
CELE_C09D8.1
Chromosome Location:
Chr.II: 10975539 - 11012254 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02406203_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ptp-3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001026771.3 NP_001021942.1 17 - 18 2917 122
NM_001026772.5 NP_001021943.1 5 - 6 870 122
NM_001267341.4 NP_001254270.1 14 - 15 2372 122
NM_001276741.3 NP_001263670.1 18 - 19 2989 122
NM_001276742.3 NP_001263671.1 18 - 19 3121 122
NM_001276743.3 NP_001263672.1 17 - 18 3049 122
NM_001377858.3 NP_001364824.1 18 - 19 2659 122
NM_001377859.2 NP_001364823.1 17 - 18 2587 122
NM_001377860.2 NP_001364658.1 15 - 16 2404 122
NM_001377861.2 NP_001364506.1 15 - 16 2112 122
NM_001377862.2 NP_001364507.1 14 - 15 2002 122

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

neuron migration
protein dephosphorylation
gastrulation
nervous system development
synapse assembly
motor neuron axon guidance
epidermis development
embryonic body morphogenesis
peptidyl-tyrosine dephosphorylation
protein localization to synapse
neuroblast migration
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
transmembrane receptor protein tyrosine phosphatase activity
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0