This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02405698_m1
See other HMR-1 GE Assays ›
Gene Symbol:
hmr-1
Gene Name:
protein-coding
Gene Aliases:
CELE_W02B9.1
Chromosome Location:
Chr.I: 11075755 - 11134667 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02405698_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: hmr-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001026479.4 NP_001021650.1 29 - 30 8134 68
NM_001264632.3 NP_001251561.1 6 - 7 2458 68
NM_001381215.1 NP_001366962.1 9 - 10 3299 68
NM_001381216.1 NP_001368105.1 4 - 5 2068 68
GenBank mRNA AJ307058.1 - 29 - 30 8134 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

establishment of mitotic spindle orientation
apical constriction involved in gastrulation
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
multicellular organism development
gastrulation
nervous system development
embryo development ending in birth or egg hatching
cell migration
cell differentiation
cortical actin cytoskeleton organization
cell migration involved in gastrulation
cell-cell adhesion
calcium ion binding
protein binding
beta-catenin binding
delta-catenin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header