This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02405368_m1
See other TOM-1 GE Assays ›
Gene Symbol:
tom-1
Gene Name:
protein-coding
Gene Aliases:
CELE_M01A10.2
Chromosome Location:
Chr.I: 5546118 - 5561689 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02405368_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: tom-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001129022.5 NP_001122494.1 7 - 8 1344 68
NM_001129023.2 NP_001122495.1 7 - 8 1347 68
NM_001313498.3 NP_001300427.1 7 - 8 1345 68
NM_001313499.3 NP_001300428.1 7 - 8 1345 68
NM_001313500.3 NP_001300429.1 7 1238 68
NM_059222.7 NP_491623.3 7 - 8 1345 68
GenBank mRNA AY912102.1 - 7 - 8 1238 68
AY997305.1 - 7 - 8 1238 68
AY997306.1 - 7 - 8 1238 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of synaptic vesicle priming
synaptic vesicle priming
regulation of exocytosis
positive regulation of GTPase activity
regulation of protein secretion
regulation of synaptic vesicle docking
regulation of presynaptic dense core granule exocytosis
cellular response to salt
GTPase activator activity
Rab GTPase binding
syntaxin binding
myosin II binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0