This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt04646865_m1
See other LOC100848574 GE Assays ›
Gene Symbol:
LOC100848574
Gene Name:
uncharacterized LOC100848574
Gene Aliases:
C1orf61, C3H1orf61
Chromosome Location:
Chr.3: 14401053 - 14425003 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt04646865_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100848574
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045650.1 4 - 5 507 78
NR_045651.1 4 - 5 481 78
NR_045652.1 4 - 5 481 78
NR_045653.1 4 - 5 506 78
NR_045654.1 4 - 5 481 78
NR_045655.1 4 - 5 550 78
NR_045656.1 2 - 3 346 78
GenBank mRNA DN534324.1 - 2 - 3 346 78
DY092185.1 - 4 - 5 309 78
DY143267.1 - 4 - 5 493 78
EE221607.1 - 4 - 5 507 78
EE228309.1 - 3 - 4 228 78
EE241358.1 - 4 - 5 333 78
EH140874.1 - 4 - 5 324 78
EV615609.1 - 4 - 5 352 78

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream myocyte enhancer factor 2D MEF2D -59.8kb Sense Chr. 3 - 14323436 - 14362406 on NCBI Build ARS-UCD1.2
Downstream Rh family B glycoprotein RHBG 17.1kb Anti-sense Chr. 3 - 14441109 - 14453884 on NCBI Build ARS-UCD1.2

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0