This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt04646846_g1
See other LOC100616098 GE Assays ›
Gene Symbol:
LOC100616098
Gene Name:
uncharacterized LOC100616098
Gene Aliases:
-
Chromosome Location:
Chr.8: 11521623 - 11522345 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt04646846_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100616098
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_038187.1 1 - 2 181 65
GenBank mRNA DT829765.1 - 1 - 2 175 65
EV739001.1 - 1 - 2 241 65

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA mir-2469 MIR2469 bta-mir-2469 Sense Chr. 8 - 11522207 - 11522273 on NCBI Build ARS-UCD1.2
Downstream DExD/H-box helicase 58 DDX58 37.3kb Anti-sense Chr. 8 - 11559690 - 11606155 on NCBI Build ARS-UCD1.2

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Bos taurus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0