This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt04646725_s1
See other MIR122 GE Assays ›
Gene Symbol:
MIR122
Gene Name:
microRNA mir-122
Gene Aliases:
MIRN122, MIRN122A, bta-mir-122, bta-mir-122a, mir-122
Chromosome Location:
Chr.24: 57588139 - 57588223 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt04646725_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR122
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_030958.1 1 39 75

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream NEDD4 like E3 ubiquitin protein ligase NEDD4L NEDD4 -37.5kb Sense Chr. 24 - 57163782 - 57550625 on NCBI Build ARS-UCD1.2
Downstream alpha kinase 2 ALPK2 23.7kb Anti-sense Chr. 24 - 57611963 - 57745147 on NCBI Build ARS-UCD1.2

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Bos taurus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0