This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt04646676_s1
See other MIR2290 GE Assays ›
Gene Symbol:
MIR2290
Gene Name:
microRNA mir-2290
Gene Aliases:
bta-mir-2290, mir-2290
Chromosome Location:
Chr.10: 12743326 - 12743405 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt04646676_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR2290
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_030835.1 1 39 64

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream DENN domain containing 4A DENND4A -53.2kb Sense Chr. 10 - 12553006 - 12690134 on NCBI Build ARS-UCD1.2
Downstream RAB11A,
member RAS oncogene family
RAB11A 240.0 Anti-sense Chr. 10 - 12743645 - 12762897 on NCBI Build ARS-UCD1.2

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Bos taurus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0