This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt04646325_mH
See other LOC503858 GE Assays ›
Gene Symbol:
LOC503858
Gene Name:
aromatase cytochrome P450 pseudogene
Gene Aliases:
-
Chromosome Location:
Chr.10: 59179025 - 59202357 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt04646325_mH
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC503858
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_003052.2 1 - 2 94 104
GenBank mRNA Z32813.1 - 95 104

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream cytochrome P450,
family 19,
subfamily A,
polypeptide 1
CYP19A1 CYP19,
CYP19P1
-22.4kb Sense Chr. 10 - 59102240 - 59156607 on NCBI Build ARS-UCD1.2
Downstream transient receptor potential cation channel subfamily M member 7 TRPM7 466.4kb Sense Chr. 10 - 59648210 - 59757063 on NCBI Build ARS-UCD1.2

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Bos taurus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0