This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt04281334_m1
See other SMIM5 GE Assays ›
Gene Symbol:
SMIM5
Gene Name:
small integral membrane protein 5
Gene Aliases:
C19H17orf109
Chromosome Location:
Chr.19: 55932583 - 55940849 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt04281334_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SMIM5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163000.1 NP_001156472.1 1 - 2 376 108
GenBank mRNA DV805795.1 - 1 - 2 376 108

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001163000.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Bos taurus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0