This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03228432_m1
See other TGFB1I1 GE Assays ›
Gene Symbol:
TGFB1I1
Gene Name:
transforming growth factor beta 1 induced transcript 1
Gene Aliases:
-
Chromosome Location:
Chr.25: 27503109 - 27509904 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03228432_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TGFB1I1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001035313.1 NP_001030390.1 6 - 7 679 68
GenBank mRNA BC104510.1 - 6 - 7 679 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of epithelial to mesenchymal transition
Wnt signaling pathway
morphogenesis of embryonic epithelium
positive regulation of transforming growth factor beta receptor signaling pathway
negative regulation of transforming growth factor beta receptor signaling pathway
ubiquitin-dependent SMAD protein catabolic process
epithelial cell differentiation
cell fate commitment
negative regulation of fat cell differentiation
positive regulation of transcription, DNA-templated
transcription coactivator activity
metal ion binding
Roundabout binding
androgen receptor binding
I-SMAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0