This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03225085_g1
See other RPL9 GE Assays ›
Gene Symbol:
RPL9
Gene Name:
ribosomal protein L9
Gene Aliases:
-
Chromosome Location:
Chr.6: 58564662 - 58569422 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03225085_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPL9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001024469.3 NP_001019640.2 1 286 85
GenBank mRNA AB099048.1 - 1 270 85
AY911321.1 - 4 - 5 292 85
BC103427.1 - 1 287 85

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC103427.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Process(es)

rRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0