This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03223687_g1
See other PRDX1 GE Assays ›
Gene Symbol:
PRDX1
Gene Name:
peroxiredoxin 1
Gene Aliases:
PRX1
Chromosome Location:
Chr.3: 100551342 - 100565167 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03223687_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRDX1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174431.1 NP_776856.1 4 - 5 423 103
GenBank mRNA AB099055.1 - 4 - 5 415 103
AF305561.1 - 4 - 5 423 103
BC148009.1 - 4 - 5 414 103
BT021073.1 - 4 - 5 411 103

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

peroxidase

Gene Ontology Categories:

Function(s)

response to oxidative stress
oxidation-reduction process
regulation of NIK/NF-kappaB signaling

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0