This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03216853_g1
See other APEX1 GE Assays ›
Gene Symbol:
APEX1
Gene Name:
apurinic/apyrimidinic endodeoxyribonuclease 1
Gene Aliases:
APEN, BAP1, REF-1
Chromosome Location:
Chr.10: 26670900 - 26673480 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03216853_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APEX1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_176609.3 NP_788782.2 4 - 5 600 61
GenBank mRNA BC122610.1 - 4 - 5 552 61
DN548789.1 - 1 - 2 65 61
DV824401.1 - 4 - 5 599 61
EH148795.1 - 4 - 5 632 61
X56685.1 - 549 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
DNA recombination
regulation of apoptotic process
regulation of mRNA stability
oxidation-reduction process
DNA demethylation
nucleic acid phosphodiester bond hydrolysis
positive regulation of nucleic acid-templated transcription
DNA binding
damaged DNA binding
transcription coactivator activity
RNA binding
DNA-(apurinic or apyrimidinic site) lyase activity
phosphoric diester hydrolase activity
3'-5' exonuclease activity
oxidoreductase activity
site-specific endodeoxyribonuclease activity, specific for altered base
chromatin DNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0