This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03216196_m1
See other CERT1 GE Assays ›
Gene Symbol:
CERT1
Gene Name:
ceramide transporter 1
Gene Aliases:
CERT, COL4A3BP
Chromosome Location:
Chr.10: 6793847 - 6925358 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03216196_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CERT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174765.3 NP_777190.1 7 - 8 1250 65
GenBank mRNA AF232931.1 - 7 - 8 1251 65
AF232933.1 - 7 - 8 1251 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
in utero embryonic development
heart morphogenesis
ceramide metabolic process
muscle contraction
endoplasmic reticulum organization
signal transduction
cell proliferation
phosphorylation
response to endoplasmic reticulum stress
lipid homeostasis
mitochondrion morphogenesis
intermembrane sphingolipid transfer
ceramide 1-phosphate transport
kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0