This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03215082_m1
See other OPN1LW GE Assays ›
Gene Symbol:
OPN1LW
Gene Name:
opsin 1, long wave sensitive
Gene Aliases:
OPSN
Chromosome Location:
Chr.X: 37321404 - 37333672 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03215082_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OPN1LW
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174566.1 NP_776991.1 3 - 4 574 83
GenBank mRNA AF280398.1 - 3 - 4 574 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s)

visual perception
detection of visible light
protein-chromophore linkage

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0