This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ba07922153_po
Gene Symbol:
Lactobacillus jensenii
Gene Name:
Pan Lactobacillus
Gene Aliases:
Lactobacillus jensenii, Lactobacillus iners, Lactobacillus gasseri, Lactobacillus crispatus
Chromosome Location:
-
Species:
Lactobacillus jensenii
Species Specific ID (Flybase ID):
-
Assay ID Ba07922153_po
Size
Availability Made To Order
Catalog #
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details


Back To Top

More Information


Updated Mapping Information

This pooled assay is composed of the following assays: Ba07922147_s1, Ba07922148_s1

Set Membership:

Microbe Detection

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0