This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ba07921923_s1
Gene Symbol:
Actinotignum schaalii
Gene Name:
Actinotignum schaalii
Gene Aliases:
Actinotignum schaalii
Chromosome Location:
-
Species:
Actinotignum schaalii
Species Specific ID (Flybase ID):
-
Assay ID Ba07921923_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details


Back To Top

More Information


Updated Mapping Information

This assay corresponds to CADT ID AIPAFMX.

Set Membership:

Recommended Assay

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0