This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ba04646155_s1
Gene Symbol:
Bacteria
Gene Name:
subclass B1 metallo-beta-lactamase VIM(blaVIM)
Gene Aliases:
Bacteria, blaVIM-1, blaVIM-10, blaVIM-11, blaVIM-12, blaVIM-13, blaVIM-14, blaVIM-15, blaVIM-16, blaVIM-17, blaVIM-18, blaVIM-19, blaVIM-2, blaVIM-20, blaVIM-23, blaVIM-24, blaVIM-25, blaVIM-26, blaVIM-27, blaVIM-28, blaVIM-29, blaVIM-3, blaVIM-30, blaVIM-31, blaVIM-32, blaVIM-33, blaVIM-34, blaVIM-35, blaVIM-36, blaVIM-37, blaVIM-38, blaVIM-39, blaVIM-4, blaVIM-40, blaVIM-41, blaVIM-42, blaVIM-43, blaVIM-44, blaVIM-45, blaVIM-46, blaVIM-47, blaVIM-48, blaVIM-49, blaVIM-5, blaVIM-50, blaVIM-51, blaVIM-52, blaVIM-53, blaVIM-54, blaVIM-55, blaVIM-56, blaVIM-57, blaVIM-58, blaVIM-59, blaVIM-6, blaVIM-60, blaVIM-62, blaVIM-63, blaVIM-64, blaVIM-65, blaVIM-66, blaVIM-67, blaVIM-68, blaVIM-70, blaVIM-72, blaVIM-73, blaVIM-74, blaVIM-75, blaVIM-76, blaVIM-77, blaVIM-78, blaVIM-79, blaVIM-8, blaVIM-9, subclass B1 metallo-beta-lactamase VIM, blaVIM
Chromosome Location:
-
Species:
Bacteria
Species Specific ID (Flybase ID):
-
Assay ID Ba04646155_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details


Back To Top

More Information


Updated Mapping Information

This assay corresponds to CADT ID AICSWV6.

Set Membership:

Recommended Assay

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0