This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ba04646152_s1
Gene Symbol:
Bacteria
Gene Name:
carbapenem-hydrolyzing class A beta-lactamase KPC(blaKPC)
Gene Aliases:
Bacteria, blaKPC-117, blaKPC-118, blaKPC-10, blaKPC-102, blaKPC-103, blaKPC-104, blaKPC-105, blaKPC-106, blaKPC-107, blaKPC-108, blaKPC-11, blaKPC-112, blaKPC-113, blaKPC-114, blaKPC-115, blaKPC-116, blaKPC-12, blaKPC-120, blaKPC-121, blaKPC-122, blaKPC-123, blaKPC-13, blaKPC-14, blaKPC-15, blaKPC-16, blaKPC-26, blaKPC-27, blaKPC-28, blaKPC-29, blaKPC-3, blaKPC-30, blaKPC-33, blaKPC-34, blaKPC-35, blaKPC-36, blaKPC-37, blaKPC-38, blaKPC-39, blaKPC-4, blaKPC-41, blaKPC-42, blaKPC-43, blaKPC-44, blaKPC-45, blaKPC-46, blaKPC-47, blaKPC-48, blaKPC-49, blaKPC-5, blaKPC-50, blaKPC-51, blaKPC-52, blaKPC-53, blaKPC-54, blaKPC-55, blaKPC-56, blaKPC-57, blaKPC-58, blaKPC-59, blaKPC-6, blaKPC-60, blaKPC-61, blaKPC-62, blaKPC-63, blaKPC-64, blaKPC-65, blaKPC-66, blaKPC-67, blaKPC-68, blaKPC-69, blaKPC-7, blaKPC-70, blaKPC-71, blaKPC-72, blaKPC-73, blaKPC-74, blaKPC-75, blaKPC-76, blaKPC-77, blaKPC-78, blaKPC-79, blaKPC-8, blaKPC-80, blaKPC-81, blaKPC-82, blaKPC-83, blaKPC-84, blaKPC-85, blaKPC-86, blaKPC-87, blaKPC-88, blaKPC-89, blaKPC-90, blaKPC-91, blaKPC-92, blaKPC-94, blaKPC-95, blaKPC-96, blaKPC-97, blaKPC-98, carbapenem-hydrolyzing class A beta-lactamase KPC, blaKPC
Chromosome Location:
-
Species:
Bacteria
Species Specific ID (Flybase ID):
-
Assay ID Ba04646152_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details


Back To Top

More Information


Set Membership:

Recommended Assay

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0