This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ba00005025_po
Gene Symbol:
Bacteria
Gene Name:
Class A extended-spectrum beta-lactamase PER
Gene Aliases:
Bacteria, blaPER-1, blaPER-10, blaPER-11, blaPER-13, blaPER-14, blaPER-15, blaPER-16, blaPER-2, blaPER-3, blaPER-4, blaPER-5, blaPER-6, blaPER-7, blaPER-8, blaPER-9, class A extended-spectrum beta-lactamase (blaPER), inhibitor-resistant class A extended-spectrum beta-lactamase (blaPER), blaPER
Chromosome Location:
-
Species:
Bacteria
Species Specific ID (Flybase ID):
-
Assay ID
Ba00005025_po
Best Coverage
Size
Availability
Made To Order
Catalog #
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Updated Mapping Information
This pooled assay is composed of the following assays: Ba04646140_s1, Ba04646157_s1
Set Membership:
Recommended Assay
Microbe Detection
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0