This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At03402525_s1
See other DUT1 GE Assays ›
Gene Symbol:
DUT1
Gene Name:
DUTP-PYROPHOSPHATASE-LIKE 1
Gene Aliases:
AT3G46940, DUTP-PYROPHOSPHATASE-LIKE 1, F13I12.6
Chromosome Location:
Chr.3: 17288134 - 17289006 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At03402525_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DUT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001339288.1 NP_001325766.1 1 502 79
NM_114561.4 NP_190278.1 1 449 79
GenBank mRNA AF370334.1 - 1 448 79
AY062989.1 - 1 363 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

hydrolase phosphatase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

dUMP biosynthetic process
DNA repair
dUTP catabolic process
magnesium ion binding
dUTP diphosphatase activity
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0