This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02356848_s1
See other AT2G07000 GE Assays ›
Gene Symbol:
AT2G07000 HEN2
Gene Name:
hypothetical protein
RNA helicase, ATP-dependent, SK12/DOB1 protein
Gene Aliases:
AT2G07000, T4E14.11, T4E14_11
AT2G06990, T4E14.10, T4E14_10, hua enhancer 2
Chromosome Location:
Chr.2: 2901870 - 2902290 on Build TAIR10.1
Chromosome Location:
Chr.2: 2894904 - 2902020 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
-
Assay ID At02356848_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT2G07000
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_126676.1 NP_178717.1 2 272 78
Gene Symbol: HEN2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_126675.3 NP_565338.1 16 4230 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
maturation of 5.8S rRNA
mRNA processing
RNA catabolic process
specification of floral organ identity
RNA metabolic process
negative regulation of posttranscriptional gene silencing
molecular_function
RNA helicase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0