This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02325290_g1
See other KT2 GE Assays ›
Gene Symbol:
KT2
Gene Name:
potassium transporter 2
Gene Aliases:
AT2G40540, ATKT2, ATKT2P, ATKUP2, KUP2, SHY3, T2P4.11, T2P4_11, TRK2, potassium transporter 2
Chromosome Location:
Chr.2: 16930555 - 16934669 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02325290_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001084563.2 NP_001078032.1 8 - 9 1977 81
NM_001336844.1 NP_001318392.1 9 - 10 1466 81
NM_001336845.1 NP_001324081.1 9 - 10 1906 81
NM_001336846.1 NP_001324082.1 9 - 10 1883 81
NM_129616.4 NP_565936.1 9 - 10 1642 81
GenBank mRNA AF012657.1 - 8 - 9 1267 81
BT004049.1 - 9 - 10 1434 81
BT005048.1 - 8 - 9 1264 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

potassium ion transport
potassium ion transmembrane transport
potassium ion transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0