This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02324873_g1
See other EIF6B GE Assays ›
Gene Symbol:
eIF6B
Gene Name:
Translation initiation factor IF6
Gene Aliases:
AT2G39820, T5I7.12, T5I7_12, eukaryotic initiation factor 6B
Chromosome Location:
Chr.2: 16618226 - 16619750 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02324873_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eIF6B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_129541.3 NP_181512.1 3 - 4 203 104

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

ribosomal subunit export from nucleus
maturation of 5.8S rRNA
maturation of LSU-rRNA
assembly of large subunit precursor of preribosome
ribosome binding
ribosomal large subunit binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0