This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02324067_g1
See other SCAR2 GE Assays ›
Gene Symbol:
SCAR2
Gene Name:
SCAR homolog 2
Gene Aliases:
AT2G38440, ATSCAR2, DIS3, IRREGULAR TRICHOME BRANCH1, IRREGULAR TRICHOME BRANCH1, ITB1, SCAR HOMOLOG 2, SCAR homolog 2, T19C21.7, T19C21_7, WAVE4
Chromosome Location:
Chr.2: 16095235 - 16101221 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02324067_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SCAR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_129400.3 NP_181378.2 3 - 4 753 93
GenBank mRNA AY743924.1 - 3 - 4 438 93
AY817016.1 - 3 - 4 659 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

trichome morphogenesis
trichome branching
actin cytoskeleton organization
actin nucleation
positive regulation of actin nucleation
positive regulation of Arp2/3 complex-mediated actin nucleation
protein binding
protein kinase A regulatory subunit binding
Arp2/3 complex binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0