This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02322889_g1
See other DML1 GE Assays ›
Gene Symbol:
DML1
Gene Name:
demeter-like 1
Gene Aliases:
AT2G36490, AtROS1, F1O11.12, F1O11_12, REPRESSOR OF SILENCING1, ROS1, demeter-like 1
Chromosome Location:
Chr.2: 15307907 - 15314807 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02322889_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DML1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_129207.5 NP_181190.3 14 - 15 3904 82
GenBank mRNA AY286009.1 - 13 - 14 3611 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
base-excision repair
DNA methylation
chromatin silencing
negative regulation of chromatin silencing
defense response to fungus
DNA demethylation
DNA-(apurinic or apyrimidinic site) lyase activity
protein binding
DNA N-glycosylase activity
class I DNA-(apurinic or apyrimidinic site) endonuclease activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0