This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02318200_g1
See other AT5G48560 GE Assays ›
Gene Symbol:
AT5G48560
Gene Name:
basic helix-loop-helix (bHLH) DNA-binding superfamily protein
Gene Aliases:
AT5G48560, CIB2, CRY2-interacting bHLH 2, K15N18.2, K15N18_2
Chromosome Location:
Chr.5: 19683809 - 19687187 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02318200_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT5G48560
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001344777.1 NP_001330141.1 5 - 6 1514 67
NM_124232.4 NP_199667.1 5 - 6 1514 67
GenBank mRNA BT002945.1 - 5 - 6 1317 67
BT005637.1 - 5 - 6 1163 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
response to blue light
transcription factor activity, sequence-specific DNA binding
protein binding
protein dimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0