This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02313035_g1
See other SIP1 GE Assays ›
Gene Symbol:
SIP1
Gene Name:
Raffinose synthase family protein
Gene Aliases:
AT5G40390, MPO12.100, MPO12_100, RS5, raffinose synthase 5, seed imbibition 1-like
Chromosome Location:
Chr.5: 16161449 - 16165453 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02313035_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SIP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_123403.4 NP_198855.1 4 - 5 2071 89
GenBank mRNA AY062781.1 - 4 - 5 1931 89
AY081645.1 - 4 - 5 1800 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

sucrose biosynthetic process
response to oxidative stress
response to water deprivation
response to abscisic acid
raffinose family oligosaccharide biosynthetic process
mannitol biosynthetic process
galactinol-sucrose galactosyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0