This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02312946_g1
See other ERA1 GE Assays ›
Gene Symbol:
ERA1
Gene Name:
Prenyltransferase family protein
Gene Aliases:
ARABIDOPSIS THALIANA FARNESYL TRANSFERASE BETA SUBUNIT, AT5G40280, ATFTB, ENHANCED RESPONSE TO ABA 1, FARNESYL TRANSFERASE BETA SUBUNIT, MSN9.180, MSN9_180, WIG, WIGGUM
Chromosome Location:
Chr.5: 16101343 - 16104962 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02312946_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ERA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001344319.1 NP_001331518.1 5 - 6 697 213
NM_123392.2 NP_198844.1 6 - 7 649 117
GenBank mRNA AF214106.1 - 6 - 7 601 117
BT030457.1 - 6 - 7 601 117

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

response to water deprivation
response to fungus
response to abscisic acid
negative regulation of abscisic acid-activated signaling pathway
regulation of meristem structural organization
protein prenylation
protein farnesylation
cuticle development
regulation of response to osmotic stress
regulation of response to salt stress
stomatal opening
farnesyltranstransferase activity
protein farnesyltransferase activity
protein binding
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0